Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYCT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025107.2 | NP_079383.2 | 2 | 808 | ||
GenBank mRNA | AF527367.1 | - | 2 | 802 | ||
AK021612.1 | - | 3 | 615 | |||
AK024922.1 | - | 2 | 800 | |||
AL834500.1 | - | 2 | 707 | |||
BC022556.1 | - | 2 | 757 | |||
GU997693.1 | - | 2 | 796 |