Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0226L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286761.1 | NP_001273690.1 | 3, 4 | 1126 | ||
NM_001286762.1 | NP_001273691.1 | 2, 3 | 1032 | |||
NM_001286763.1 | NP_001273692.1 | 3, 4 | 761 | |||
NM_001286764.1 | NP_001273693.1 | 2, 3 | 645 | |||
NM_025113.3 | NP_079389.2 | 3, 4 | 1148 | |||
GenBank mRNA | AK025215.1 | - | 2, 3 | 781 | ||
AK093073.1 | - | 3, 4 | 1096 | |||
AK293034.1 | - | 2, 3 | 397 | |||
AK310264.1 | - | 4, 5 | 1135 | |||
BC032311.1 | - | 2, 3 | 713 | |||
BC043488.1 | - | 3, 4 | 494 | 5205680 | ||
BX401854.2 | - | 3, 4 | 881 | |||
BX641126.1 | - | 1, 2 | 182 |