Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025145.5 | NP_079421.5 | 2 | 408 | ||
XM_005270171.2 | XP_005270228.1 | 2 | 409 | |||
XM_005270172.3 | XP_005270229.1 | 2 | 409 | |||
XM_011540196.2 | XP_011538498.1 | 2 | 409 | |||
XM_011540197.2 | XP_011538499.1 | 2 | 409 | |||
XM_011540198.2 | XP_011538500.1 | 2 | 409 | |||
XM_011540199.2 | XP_011538501.1 | 2 | 409 | |||
XM_011540200.2 | XP_011538502.1 | 2 | 409 | |||
XM_011540201.2 | XP_011538503.1 | 2 | 409 | |||
XM_017016681.1 | XP_016872170.1 | 2 | 409 | |||
XM_017016682.1 | XP_016872171.1 | 2 | 409 | |||
XM_017016684.1 | XP_016872173.1 | 2 | 409 | |||
GenBank mRNA | AK026597.1 | - | 2 | 386 | ||
AK295018.1 | - | 2 | 387 | |||
AL136901.1 | - | 2 | 350 | |||
BC036647.2 | - | 2 | 408 | 4825754 | ||
DB065957.1 | - | 2 | 412 |