Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2G2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202489.1 | NP_001189418.1 | 4 | 745 | ||
NM_003343.5 | NP_003334.2 | 6 | 827 | |||
NM_182688.2 | NP_872630.1 | 7 | 942 | |||
XM_017028469.1 | XP_016883958.1 | 6 | 786 | |||
GenBank mRNA | AF032456.1 | - | 6 | 794 | ||
AK075143.1 | - | 2 | 968 | |||
AK122700.1 | - | 2 | 2244 | |||
AK290629.1 | - | 6 | 826 | |||
AL834129.1 | - | 2 | 1561 | |||
BC001738.1 | - | 6 | 790 | 3536853 | ||
BC008351.1 | - | 6 | 787 | 3536171 | ||
BC011569.1 | - | 6 | 799 | 4548763 |