Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115113.2 | NP_001108585.2 | 6 | 1092 | ||
NM_025160.6 | NP_079436.4 | 6 | 1140 | |||
XM_005273273.4 | XP_005273330.1 | 6 | 1041 | |||
XM_006711817.3 | XP_006711880.1 | 6 | 1041 | |||
XM_011544281.2 | XP_011542583.1 | 6 | 881 | |||
XM_011544282.2 | XP_011542584.1 | 3 | 203 | |||
XM_017002396.1 | XP_016857885.1 | 6 | 729 | |||
GenBank mRNA | AB209887.1 | - | 6 | 1140 | ||
AB586698.1 | - | 1 | 49 | |||
AK024669.1 | - | 2 | 154 | |||
AK225061.1 | - | 2 | 154 | |||
AK299230.1 | - | 3 | 325 | |||
AK307909.1 | - | 6 | 806 | |||
AY221751.1 | - | 6 | 930 | |||
AY304473.1 | - | 6 | 697 | |||
BC031471.2 | - | 1 | 83 | |||
BC034498.1 | - | 6 | 943 | |||
BC052301.1 | - | 5 | 536 | 6420398 | ||
BC063817.1 | - | 2 | 202 | |||
EF011612.1 | - | 5 | 535 |