Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115113.2 | NP_001108585.2 | 7 | 1198 | ||
NM_025160.6 | NP_079436.4 | 7 | 1246 | |||
XM_005273273.4 | XP_005273330.1 | 7 | 1147 | |||
XM_006711817.3 | XP_006711880.1 | 7 | 1144 | |||
XM_011544281.2 | XP_011542583.1 | 7 | 987 | |||
XM_011544282.2 | XP_011542584.1 | 4 | 309 | |||
XM_017002396.1 | XP_016857885.1 | 7 | 835 | |||
GenBank mRNA | AB209887.1 | - | 7 | 1246 | ||
AK024669.1 | - | 3 | 260 | |||
AK225061.1 | - | 3 | 260 | |||
AK299230.1 | - | 4 | 431 | |||
AY221751.1 | - | 7 | 1036 | |||
AY304473.1 | - | 7 | 803 | |||
BC031471.2 | - | 2 | 189 | |||
BC034498.1 | - | 8 | 1132 | |||
BC052301.1 | - | 6 | 642 | 6420398 | ||
BC063817.1 | - | 3 | 308 | |||
EF011612.1 | - | 6 | 641 |