Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPKC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025194.2 | NP_079470.1 | 7 | 2106 | ||
XM_017027324.1 | XP_016882813.1 | 8 | 1290 | |||
GenBank mRNA | AK024596.1 | - | 7 | 1141 | ||
BC060788.1 | - | 7 | 2106 | 30332196 | ||
D38169.1 | - | 7 | 1837 |