Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272004.1 | NP_001258933.1 | 1 | 398 | ||
NM_025209.3 | NP_079485.1 | 1 | 398 | |||
NR_104159.1 | 1 | 398 | ||||
NR_104160.1 | 1 | 398 | ||||
GenBank mRNA | AF277374.1 | - | 1 | 365 | ||
AK309822.1 | - | 1 | 267 | |||
BC036529.1 | - | 1 | 399 | 4828057 | ||
BG719217.1 | - | 1 | 306 | |||
BI461563.1 | - | 1 | 399 | |||
BX476993.1 | - | 1 | 389 | |||
DA333251.1 | - | 1 | 382 | |||
DB447845.1 | - | 1 | 336 |