Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD1LG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025239.3 | NP_079515.2 | 7 | 1130 | ||
XM_005251600.3 | XP_005251657.1 | 7 | 1241 | |||
GenBank mRNA | AF329193.1 | - | 7 | 1115 | ||
AF344424.1 | - | 7 | 1130 | |||
AK001872.1 | - | 5 | 561 | |||
BC113678.1 | - | 7 | 893 | 8322730 | ||
BC113680.1 | - | 7 | 893 | 8322732 | ||
BC143920.1 | - | 7 | 897 |