Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025245.2 | NP_079521.1 | 8 | 1472 | ||
NR_038198.1 | 8 | 1371 | ||||
XM_006722911.3 | XP_006722974.1 | 7 | 1053 | |||
XM_011528320.2 | XP_011526622.1 | 9 | 1518 | |||
XM_011528322.2 | XP_011526624.1 | 7 | 1107 | |||
XM_017027326.1 | XP_016882815.1 | 9 | 1153 | |||
XM_017027327.1 | XP_016882816.1 | 9 | 1134 | |||
XM_017027329.1 | XP_016882818.1 | 6 | 943 | |||
GenBank mRNA | AJ300182.1 | - | 7 | 1053 | ||
AK097427.1 | - | 8 | 1209 | |||
BC141859.1 | - | 8 | 1472 | 8860384 |