Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPK3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030570.3 | NP_085047.1 | 3 | 913 | ||
NM_182683.2 | NP_872624.1 | 4 | 748 | |||
NM_182684.2 | NP_872625.1 | 5 | 828 | |||
GenBank mRNA | AK056760.1 | - | 4 | 556 | ||
AK290096.1 | - | 3 | 816 | |||
AY233462.1 | - | 5 | 661 | |||
BC004304.1 | - | 3 | 737 | 3625247 |
Gene Symbol: LOC105375355 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005250612.3 | XP_005250669.1 | 3 | 1234 | ||
XM_006716140.3 | XP_006716203.1 | 5 | 906 | |||
XM_006716141.3 | XP_006716204.1 | 4 | 826 |