Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPEB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079533.1 | NP_001073001.1 | 11 | 1951 | ||
NM_001079534.1 | NP_001073002.1 | 11 | 1936 | |||
NM_001079535.1 | NP_001073003.1 | 10 | 2037 | |||
NM_001288819.1 | NP_001275748.1 | 12 | 2390 | |||
NM_001288820.1 | NP_001275749.1 | 10 | 1709 | |||
NM_030594.4 | NP_085097.3 | 12 | 2082 | |||
GenBank mRNA | AF329402.1 | - | 12 | 2268 | ||
AF329403.1 | - | 11 | 1934 | |||
AK023265.1 | - | 11 | 1876 | |||
AK294280.1 | - | 11 | 1962 | |||
AK300105.1 | - | 12 | 2390 | |||
AL832156.1 | - | 8 | 1744 | |||
BC035348.1 | - | 10 | 1964 | 5195793 | ||
BC050629.1 | - | 12 | 2043 | 6047179 | ||
BX649159.1 | - | 13 | 3822 |