Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LNPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305008.1 | NP_001291937.1 | 13 | 1861 | ||
NM_001305009.1 | NP_001291938.1 | 14 | 1949 | |||
NM_001305010.1 | NP_001291939.1 | 12 | 1669 | |||
NM_001305011.1 | NP_001291940.1 | 11 | 1626 | |||
NM_030650.2 | NP_085153.1 | 13 | 1856 | |||
NR_130941.1 | 15 | 2220 | ||||
XM_005246878.3 | XP_005246935.1 | 14 | 1875 | |||
XM_005246879.4 | XP_005246936.1 | 13 | 1803 | |||
XM_005246880.3 | XP_005246937.1 | 9 | 1493 | |||
XM_006712783.2 | XP_006712846.1 | 13 | 1998 | |||
XM_011511945.2 | XP_011510247.1 | 14 | 2207 | |||
XM_017005055.1 | XP_016860544.1 | 9 | 1401 | |||
GenBank mRNA | AB051502.1 | - | 13 | 1763 | ||
AK056532.1 | - | 13 | 1779 | |||
AL832947.1 | - | 10 | 1519 | |||
BC031530.1 | - | 2 | 709 | |||
BC110329.1 | - | 11 | 1555 | 5732809 | ||
CR936742.1 | - | 13 | 1805 |