Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPH6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030785.3 | NP_110412.1 | 1 | 668 | ||
XM_011527351.1 | XP_011525653.1 | 1 | 644 | |||
GenBank mRNA | AK098837.1 | - | 1 | 613 | ||
AK223347.1 | - | 1 | 613 | |||
AL136761.1 | - | 1 | 640 | |||
BC057785.1 | - | 1 | 496 | |||
BC136351.1 | - | 1 | 549 | 9020338 | ||
BC136352.1 | - | 1 | 549 | 9020339 |