Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMAN2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142292.1 | NP_001135764.1 | 9 | 2284 | ||
NM_001322346.1 | NP_001309275.1 | 8 | 2217 | |||
NM_001322347.1 | NP_001309276.1 | 8 | 2181 | |||
NM_001322350.1 | NP_001309279.1 | 7 | 2148 | |||
NM_001322351.1 | NP_001309280.1 | 6 | 2065 | |||
NM_001322352.1 | NP_001309281.1 | 10 | 2381 | |||
NM_001322354.1 | NP_001309283.1 | 8 | 2231 | |||
NM_001322355.1 | NP_001309284.1 | 8 | 2265 | |||
NM_001322356.1 | NP_001309285.1 | 7 | 2184 | |||
NM_030805.3 | NP_110432.1 | 8 | 2251 | |||
GenBank mRNA | AK075347.1 | - | 8 | 2239 | ||
AL136617.1 | - | 8 | 2242 | |||
AY358929.1 | - | 8 | 2222 | |||
BC000347.2 | - | 8 | 2214 | |||
BC004505.2 | - | 1 | 632 | |||
BC005822.2 | - | 8 | 2214 | |||
BC005862.2 | - | 8 | 2214 | |||
BC067265.1 | - | 8 | 2221 | 6502212 | ||
BX648002.1 | - | 9 | 2262 |