Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024938.3 | NP_001020109.1 | 12 | 1652 | ||
NM_001024939.3 | NP_001020110.1 | 12 | 1540 | |||
NM_001282864.1 | NP_001269793.1 | 11 | 1894 | |||
NM_030807.4 | NP_110434.3 | 13 | 1763 | |||
NR_104247.1 | 11 | 1499 | ||||
GenBank mRNA | AB209301.1 | - | 8 | 2125 | ||
AF443201.1 | - | 13 | 1594 | |||
AJ271290.1 | - | 12 | 1483 | |||
AK075467.1 | - | 12 | 1428 | |||
AK092466.1 | - | 2 | 1564 | |||
AK300196.1 | - | 11 | 1893 | |||
AK314502.1 | - | 13 | 1534 | |||
BC018967.2 | - | 1 | 208 | |||
BC094735.1 | - | 12 | 1469 | 5304025 | ||
BC100807.2 | - | 9 | 1601 | |||
BC100808.2 | - | 12 | 1398 | 40000757 | ||
BC100809.1 | - | 12 | 1398 | 40000760 | ||
BC100810.1 | - | 11 | 1245 | |||
CR456373.1 | - | 12 | 1429 |