Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL21A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318751.1 | NP_001305680.1 | 7 | 1364 | ||
NM_001318752.1 | NP_001305681.1 | 6 | 1254 | |||
NM_030820.3 | NP_110447.2 | 6 | 1290 | |||
NR_134849.1 | 6 | 1290 | ||||
NR_134850.1 | 6 | 1290 | ||||
NR_134851.1 | 6 | 1290 | ||||
XM_006715223.1 | XP_006715286.1 | 6 | 1456 | |||
XM_011514924.2 | XP_011513226.1 | 6 | 1233 | |||
XM_011514925.2 | XP_011513227.1 | 8 | 1403 | |||
XM_011514926.1 | XP_011513228.1 | 6 | 1161 | |||
XM_011514927.1 | XP_011513229.1 | 6 | 1115 | |||
GenBank mRNA | AF330693.1 | - | 6 | 1220 | ||
AF414088.1 | - | 6 | 1260 | |||
AK126586.1 | - | 6 | 1157 | |||
AK313398.1 | - | 6 | 1266 | |||
AL136624.1 | - | 6 | 1260 | |||
BC045597.1 | - | 6 | 1245 | 4814199 | ||
BC126108.1 | - | 6 | 1145 | 8991824 | ||
BC143862.1 | - | 6 | 1145 | |||
BC143863.1 | - | 6 | 1145 | |||
BC143864.1 | - | 6 | 1145 | |||
BC143865.1 | - | 7 | 1219 | 9052381 |