Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR5V1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030876.5 | NP_110503.3 | 1 | 233 | ||
XM_011514929.2 | XP_011513231.1 | 3 | 408 | |||
XM_011514930.2 | XP_011513232.1 | 2 | 286 | |||
GenBank mRNA | AJ459859.1 | - | 1 | 236 | ||
BC069149.1 | - | 1 | 151 |