Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003365.2 | NP_003356.2 | 3, 4 | 350 | ||
GenBank mRNA | AK129903.1 | - | 3, 4 | 330 | ||
AK300694.1 | - | 3, 4 | 285 | |||
AK302840.1 | - | 3, 4 | 375 | |||
AK313090.1 | - | 3, 4 | 320 | |||
BC009586.1 | - | 3, 4 | 321 | 4096628 | ||
D26485.1 | - | 3, 4 | 312 | |||
L16842.1 | - | 3, 4 | 722 |