Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030918.5 | NP_112180.4 | 12 | 6689 | ||
XM_005245509.2 | XP_005245566.1 | 12 | 6034 | |||
XM_005245510.3 | XP_005245567.1 | 12 | 5644 | |||
XM_005245511.4 | XP_005245568.1 | 12 | 6717 | |||
XM_011510024.2 | XP_011508326.1 | 11 | 5755 | |||
XM_011510025.2 | XP_011508327.1 | 11 | 5613 | |||
XM_017002417.1 | XP_016857906.1 | 11 | 6390 | |||
GenBank mRNA | AB007957.2 | - | 12 | 6664 | ||
AK001395.1 | - | 1 | 2451 |