Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAZALD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030929.4 | NP_112191.2 | 5 | 1997 | ||
GenBank mRNA | AK172862.1 | - | 5 | 1248 | ||
AK225949.1 | - | 5 | 1249 | |||
AY359087.1 | - | 5 | 1253 | |||
BC007758.2 | - | 5 | 1398 | 4122366 | ||
BI766345.1 | - | 4 | 709 |