Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIAPH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042517.1 | NP_001035982.1 | 19 | 2469 | ||
NM_001258366.1 | NP_001245295.1 | 18 | 2436 | |||
NM_001258367.1 | NP_001245296.1 | 17 | 2331 | |||
NM_001258368.1 | NP_001245297.1 | 17 | 2259 | |||
NM_001258369.1 | NP_001245298.1 | 19 | 2469 | |||
NM_001258370.1 | NP_001245299.1 | 13 | 1640 | |||
NM_030932.3 | NP_112194.2 | 13 | 1640 | |||
XM_006719876.1 | XP_006719939.1 | 13 | 1620 | |||
XM_011535258.2 | XP_011533560.1 | 19 | 2412 | |||
GenBank mRNA | AB244756.1 | - | 18 | 2217 | ||
AB244757.1 | - | 17 | 2112 | |||
AB244758.1 | - | 17 | 2040 | |||
AK298509.1 | - | 19 | 2255 | |||
AK307652.1 | - | 13 | 1620 | |||
AK316474.1 | - | 19 | 2382 | |||
AL137718.1 | - | 13 | 1612 | |||
AY750055.1 | - | 19 | 2342 | |||
AY818645.1 | - | 19 | 2250 | |||
BC034952.1 | - | 13 | 1644 | 4830888 | ||
BC048963.1 | - | 13 | 1596 | 5265490 | ||
BC068504.1 | - | 13 | 1596 | 4838507 | ||
BG203073.1 | - | 2 | 225 |