Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIAPH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042517.1 | NP_001035982.1 | 21 | 2803 | ||
NM_001258366.1 | NP_001245295.1 | 20 | 2770 | |||
NM_001258367.1 | NP_001245296.1 | 19 | 2665 | |||
NM_001258368.1 | NP_001245297.1 | 19 | 2593 | |||
NM_001258369.1 | NP_001245298.1 | 21 | 2803 | |||
NM_001258370.1 | NP_001245299.1 | 15 | 1974 | |||
NM_030932.3 | NP_112194.2 | 15 | 1974 | |||
XM_006719876.1 | XP_006719939.1 | 15 | 1954 | |||
XM_011535258.2 | XP_011533560.1 | 21 | 2746 | |||
GenBank mRNA | AB244756.1 | - | 20 | 2551 | ||
AB244757.1 | - | 19 | 2446 | |||
AB244758.1 | - | 19 | 2374 | |||
AK298509.1 | - | 21 | 2589 | |||
AK307652.1 | - | 15 | 1954 | |||
AK316474.1 | - | 21 | 2716 | |||
AL137718.1 | - | 15 | 1946 | |||
AY750055.1 | - | 21 | 2676 | |||
AY818645.1 | - | 21 | 2584 | |||
BC034952.1 | - | 15 | 1978 | 4830888 | ||
BC048963.1 | - | 15 | 1930 | 5265490 | ||
BC068504.1 | - | 15 | 1930 | 4838507 |