Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320153.1 | NP_001307082.1 | 10 | 1788 | ||
NM_001320155.1 | NP_001307084.1 | 10 | 1078 | |||
NM_030937.4 | NP_112199.2 | 9 | 1064 | |||
NR_135154.1 | 11 | 3218 | ||||
XM_011542216.2 | XP_011540518.1 | 9 | 1139 | |||
XM_011542219.2 | XP_011540521.1 | 9 | 948 | |||
XM_011542221.2 | XP_011540523.1 | 8 | 779 | |||
XM_017002420.1 | XP_016857909.1 | 10 | 2569 | |||
XM_017002421.1 | XP_016857910.1 | 11 | 1452 | |||
XM_017002422.1 | XP_016857911.1 | 10 | 1186 | |||
GenBank mRNA | AF087903.1 | - | 2 | 108 | ||
AF251294.1 | - | 10 | 1667 | |||
AK000685.1 | - | 6 | 741 | |||
AK026796.1 | - | 5 | 1361 | |||
AK027770.1 | - | 4 | 2033 | |||
AK056120.1 | - | 10 | 1760 | |||
AK057133.1 | - | 8 | 779 | |||
AK074112.1 | - | 11 | 3189 | |||
AK225758.1 | - | 9 | 1059 | |||
AK292268.1 | - | 4 | 587 | |||
AK293451.1 | - | 6 | 542 | |||
AY037150.1 | - | 9 | 1038 | |||
AY116620.1 | - | 10 | 1067 | |||
BC071622.1 | - | 4 | 451 |