Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030940.3 | NP_112202.2 | 4 | 398 | ||
GenBank mRNA | AF284752.1 | - | 4 | 353 | ||
AK055643.1 | - | 4 | 361 | |||
AK225937.1 | - | 4 | 361 | |||
AK296093.1 | - | 4 | 584 | |||
AL834356.1 | - | 4 | 343 | |||
AY015642.1 | - | 4 | 333 | |||
BC002675.2 | - | 4 | 352 | 3608301 | ||
BC053900.1 | - | 4 | 319 | |||
BC071621.1 | - | 4 | 332 | 5107151 | ||
BG772866.1 | - | 4 | 399 |