Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PITPNM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165966.1 | NP_001159438.1 | 9 | 1148 | ||
NM_031220.3 | NP_112497.2 | 10 | 1256 | |||
XM_011524015.2 | XP_011522317.1 | 10 | 1196 | |||
XM_011524016.2 | XP_011522318.1 | 10 | 1197 | |||
XM_011524017.2 | XP_011522319.1 | 10 | 1198 | |||
GenBank mRNA | AB209130.1 | - | 4 | 3619 | ||
AF334586.1 | - | 10 | 1253 | |||
AL389994.1 | - | 9 | 979 | |||
BC128583.1 | - | 10 | 1193 | 40128716 | ||
BC128584.1 | - | 10 | 1193 | 40128724 |