Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PITPNM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165966.1 | NP_001159438.1 | 12 | 1697 | ||
NM_031220.3 | NP_112497.2 | 13 | 1805 | |||
XM_011524015.2 | XP_011522317.1 | 13 | 1745 | |||
XM_011524016.2 | XP_011522318.1 | 13 | 1746 | |||
XM_011524017.2 | XP_011522319.1 | 13 | 1756 | |||
GenBank mRNA | AB209130.1 | - | 7 | 4168 | ||
AF334586.1 | - | 13 | 1802 | |||
AL389994.1 | - | 12 | 1528 | |||
BC020484.1 | - | 4 | 403 | |||
BC128583.1 | - | 13 | 1742 | 40128716 | ||
BC128584.1 | - | 13 | 1742 | 40128724 |