Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031280.3 | NP_112570.2 | 7, 8 | 793 | ||
GenBank mRNA | AB049946.1 | - | 7, 8 | 668 | ||
AB209905.1 | - | 5, 6 | 4946 | |||
AF265439.1 | - | 7, 8 | 712 | |||
AK021626.1 | - | 3, 4 | 1453 | |||
AK125230.1 | - | 2, 3 | 1967 | |||
AK315441.1 | - | 7, 8 | 732 | |||
BC031336.1 | - | 7, 8 | 708 | 3543896 | ||
BM048518.1 | - | 7 | 793 | |||
BU927793.1 | - | 7, 8 | 643 | |||
CF123360.1 | - | 1, 2 | 198 |