Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313726.1 | NP_001300655.1 | 21 | 2243 | ||
NM_001313727.1 | NP_001300656.1 | 17 | 1801 | |||
NM_031418.3 | NP_113606.2 | 20 | 2151 | |||
XM_011520282.2 | XP_011518584.1 | 7 | 698 | |||
XM_017018118.1 | XP_016873607.1 | 18 | 1779 | |||
XM_017018119.1 | XP_016873608.1 | 15 | 1391 | |||
GenBank mRNA | AJ300461.1 | - | 20 | 2858 | ||
AK295143.1 | - | 17 | 1713 | |||
AK295658.1 | - | 4 | 186 | |||
AK295816.1 | - | 17 | 1713 | |||
AK302659.1 | - | 20 | 2190 | |||
AK304781.1 | - | 21 | 2084 | |||
AK316018.1 | - | 17 | 1713 | |||
BQ430769.1 | - | 1 | 22 |