Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIF1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185095.1 | NP_001172024.1 | 7 | 1834 | ||
NM_001185096.1 | NP_001172025.1 | 6 | 1685 | |||
NM_031426.3 | NP_113614.1 | 6 | 1756 | |||
NR_033701.1 | 6 | 1778 | ||||
XM_017015177.1 | XP_016870666.1 | 5 | 1690 | |||
GenBank mRNA | AK022845.1 | - | 7 | 1701 | ||
AK027181.1 | - | 1 | 151 | |||
AK091912.1 | - | 6 | 1670 | |||
AK128526.1 | - | 7 | 1779 | |||
AL136566.1 | - | 6 | 1690 | |||
AL833896.1 | - | 6 | 1690 | |||
BC021253.2 | - | 6 | 1611 | 3504509 |