Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031431.3 | NP_113619.2 | 12 | 1382 | ||
XM_011535266.1 | XP_011533568.1 | 12 | 1189 | |||
XM_011535267.2 | XP_011533569.1 | 12 | 1362 | |||
XM_017020790.1 | XP_016876279.1 | 11 | 1035 | |||
GenBank mRNA | AF332595.1 | - | 12 | 1341 | ||
AF349676.1 | - | 12 | 1361 | |||
AK026305.1 | - | 12 | 1341 | |||
AK225466.1 | - | 12 | 1341 | |||
AK225952.1 | - | 12 | 1341 | |||
AK304248.1 | - | 12 | 1184 | |||
AK314387.1 | - | 12 | 1308 | |||
BC038953.1 | - | 12 | 1272 | 5756412 | ||
BX648211.1 | - | 12 | 945 |