Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003385.4 | NP_003376.2 | 2 | 517 | ||
GenBank mRNA | AB001104.1 | - | 2 | 272 | ||
AF039555.1 | - | 2 | 325 | |||
AK308816.1 | - | 383 | ||||
BC022012.2 | - | 2 | 354 | 4823954 | ||
BI544338.1 | - | 2 | 518 | |||
U14747.1 | - | 2 | 340 |