Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031457.1 | NP_113645.1 | 7 | 1039 | ||
GenBank mRNA | AF237905.1 | - | 7 | 1039 | ||
BC022895.1 | - | 7 | 964 | 4691480 |