Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD99L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184808.1 | NP_001171737.1 | 7 | 683 | ||
NM_001242614.1 | NP_001229543.1 | 11 | 932 | |||
NM_031462.3 | NP_113650.2 | 10 | 902 | |||
NM_134445.3 | NP_604394.1 | 7 | 686 | |||
NM_134446.3 | NP_604395.1 | 8 | 755 | |||
GenBank mRNA | AK023776.1 | - | 9 | 588 | ||
AK024278.1 | - | 6 | 409 | |||
AK125020.1 | - | 11 | 743 | |||
AK126859.1 | - | 1 | 1301 | |||
AK290200.1 | - | 10 | 678 | |||
AK291375.1 | - | 8 | 568 | |||
AK293244.1 | - | 7 | 489 | |||
AL136580.1 | - | 10 | 791 | |||
AY078165.1 | - | 10 | 710 | |||
AY078166.1 | - | 8 | 563 | |||
AY078167.1 | - | 7 | 494 | |||
AY358837.1 | - | 10 | 775 | |||
BC025729.1 | - | 11 | 2123 | 5203087 | ||
BC030536.1 | - | 10 | 762 | 5242536 | ||
KU178797.1 | - | 10 | 674 | |||
KU178798.1 | - | 4 | 245 |