Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GORASP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278789.1 | NP_001265718.1 | 7 | 2569 | ||
NM_001278790.1 | NP_001265719.1 | 6 | 2389 | |||
NM_031899.3 | NP_114105.1 | 9 | 3317 | |||
NR_103866.1 | 9 | 2813 | ||||
NR_103867.1 | 9 | 2805 | ||||
XM_006713301.3 | XP_006713364.1 | 9 | 3417 | |||
XM_011534020.2 | XP_011532322.1 | 9 | 3327 | |||
XM_011534021.2 | XP_011532323.1 | 8 | 3237 | |||
XM_017007050.1 | XP_016862539.1 | 9 | 2503 | |||
XM_017007051.1 | XP_016862540.1 | 8 | 2852 | |||
XM_017007052.1 | XP_016862541.1 | 8 | 2448 | |||
GenBank mRNA | AJ409349.1 | - | 9 | 2593 | ||
AK001574.1 | - | 8 | 2835 | |||
AK027096.1 | - | 857 | ||||
AK057043.1 | - | 6 | 2127 | |||
AK124755.1 | - | 9 | 3317 | |||
BC008928.2 | - | 9 | 2534 | |||
BC075854.1 | - | 9 | 2603 | 5503263 | ||
BC103826.1 | - | 9 | 2535 | 40001584 | ||
BC103827.1 | - | 9 | 2535 | 40001588 |