Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031903.2 | NP_114109.1 | 2 | 312 | ||
GenBank mRNA | AB049649.1 | - | 2 | 311 | ||
AF161401.1 | - | 2 | 252 | |||
AK090551.1 | - | 2 | 276 | |||
AK313393.1 | - | 2 | 270 | |||
BC013147.1 | - | 2 | 271 | 4340082 | ||
BM833110.1 | - | 2 | 275 | |||
DB456673.1 | - | 2 | 454 |