Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031914.2 | NP_114120.2 | 6 | 1998 | ||
XM_006720272.3 | XP_006720335.1 | 8 | 2458 | |||
XM_011537219.1 | XP_011535521.1 | 8 | 2535 | |||
XM_011537223.2 | XP_011535525.1 | 7 | 1572 | |||
XM_011537224.2 | XP_011535526.1 | 8 | 1718 | |||
XM_011537225.2 | XP_011535527.1 | 8 | 2017 | |||
XM_011537227.2 | XP_011535529.2 | 6 | 1395 | |||
XM_011537228.2 | XP_011535530.1 | 6 | 1338 | |||
XM_011537229.2 | XP_011535531.1 | 5 | 1182 | |||
XM_017021693.1 | XP_016877182.1 | 8 | 2535 | |||
XM_017021694.1 | XP_016877183.1 | 8 | 2224 | |||
XM_017021695.1 | XP_016877184.1 | 9 | 2369 | |||
XM_017021696.1 | XP_016877185.1 | 7 | 1517 | |||
XM_017021698.1 | XP_016877187.1 | 7 | 1641 | |||
GenBank mRNA | AB102949.1 | - | 6 | 1741 | ||
AJ303367.1 | - | 1 | 152 | |||
AJ617628.1 | - | 6 | 1999 | |||
BC040924.1 | - | 4 | 890 | |||
BC117363.1 | - | 6 | 1857 | 40125914 | ||
BC143598.1 | - | 6 | 1857 | 9052110 |