Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSD1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145313.2 | NP_001138785.1 | 4 | 504 | ||
NM_001287191.1 | NP_001274120.1 | 3 | 408 | |||
NM_001287192.1 | NP_001274121.1 | 3 | 408 | |||
NM_031919.4 | NP_114125.1 | 3 | 408 | |||
XM_005252254.2 | XP_005252311.1 | 4 | 737 | |||
XM_005252257.3 | XP_005252314.1 | 3 | 403 | |||
XM_005252260.3 | XP_005252317.1 | 3 | 393 | |||
XM_011519077.2 | XP_011517379.1 | 5 | 395 | |||
XM_011519078.2 | XP_011517380.2 | 4 | 402 | |||
XM_011519079.2 | XP_011517381.2 | 4 | 404 | |||
XM_017015182.1 | XP_016870671.1 | 4 | 737 | |||
XM_017015183.1 | XP_016870672.1 | 3 | 403 | |||
XM_017015184.1 | XP_016870673.1 | 1 | 38 | |||
XM_017015185.1 | XP_016870674.1 | 4 | 410 | |||
XM_017015187.1 | XP_016870676.1 | 3 | 403 | |||
GenBank mRNA | AF088031.1 | - | 3 | 397 | ||
AF316830.1 | - | 3 | 271 | |||
AK296433.1 | - | 3 | 319 | |||
AK299350.1 | - | 4 | 436 | |||
AK299491.1 | - | 3 | 319 | |||
AK309817.1 | - | 4 | 745 | |||
BP219536.1 | - | 4 | 476 | |||
BP229804.1 | - | 3 | 408 | |||
BP365251.1 | - | 3 | 398 | |||
DN999402.1 | - | 3 | 251 |