Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031936.4 | NP_114142.3 | 2 | 1671 | ||
XM_017002438.1 | XP_016857927.1 | 2 | 4004 | |||
GenBank mRNA | AF258342.1 | - | 3 | 1307 | ||
AF317652.1 | - | 1 | 988 | |||
AK290027.1 | - | 2 | 1671 | |||
BC067464.1 | - | 1 | 1056 | 6971762 | ||
JF810889.1 | - | 1 | 988 |
Set Membership: |
G-protein coupled receptor |