Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320458.1 | NP_001307387.1 | 4 | 769 | ||
NM_003396.2 | NP_003387.1 | 4 | 769 | |||
XM_011525178.2 | XP_011523480.1 | 4 | 879 | |||
GenBank mRNA | AB063483.1 | - | 4 | 753 | ||
AK127268.1 | - | 4 | 790 | |||
AK127615.1 | - | 4 | 769 | |||
AY358217.1 | - | 4 | 789 | |||
BC064534.1 | - | 4 | 743 | 5588904 |