Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317781.1 | NP_001304710.1 | 5 | 806 | ||
XM_005263267.4 | XP_005263324.1 | 5 | 1249 | |||
XM_011532302.2 | XP_011530604.1 | 5 | 1249 | |||
XM_011532303.2 | XP_011530605.1 | 5 | 1249 | |||
XM_011532304.2 | XP_011530606.1 | 5 | 1249 | |||
XM_011532305.2 | XP_011530607.1 | 5 | 1249 | |||
XM_011532306.2 | XP_011530608.1 | 5 | 1249 | |||
XM_011532307.2 | XP_011530609.1 | 5 | 1249 | |||
XM_011532308.2 | XP_011530610.1 | 5 | 1249 | |||
XM_017008680.1 | XP_016864169.1 | 5 | 1249 | |||
XM_017008681.1 | XP_016864170.1 | 5 | 1249 | |||
XM_017008682.1 | XP_016864171.1 | 5 | 1249 | |||
XM_017008683.1 | XP_016864172.1 | 2 | 158 | |||
XM_017008684.1 | XP_016864173.1 | 5 | 529 | |||
XM_017008686.1 | XP_016864175.1 | 5 | 1249 | |||
XM_017008687.1 | XP_016864176.1 | 5 | 1249 | |||
GenBank mRNA | AK095661.1 | - | 2 | 210 | ||
AY601647.1 | - | 5 | 806 | |||
BM554003.1 | - | 5 | 807 | |||
BX508898.1 | - | 4 | 532 |