Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006713783.3 | XP_006713846.1 | 7 | 2912 | ||
XM_017007311.1 | XP_016862800.1 | 10 | 3147 | |||
XM_017007312.1 | XP_016862801.1 | 8 | 3146 | |||
GenBank mRNA | BU183949.1 | - | 609 |