Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042403.2 | NP_001035862.1 | 2 | 942 | ||
NM_001278393.1 | NP_001265322.1 | 2 | 1158 | |||
NM_032147.4 | NP_115523.2 | 2 | 1087 | |||
NR_125360.1 | 1 | 868 | ||||
XM_005269172.1 | XP_005269229.1 | 2 | 1081 | |||
XM_005269173.1 | XP_005269230.1 | 2 | 1158 | |||
XM_005269174.1 | XP_005269231.1 | 2 | 925 | |||
XM_011538800.2 | XP_011537102.1 | 3 | 1108 | |||
XM_011538801.1 | XP_011537103.1 | 3 | 968 | |||
XM_011538802.1 | XP_011537104.1 | 2 | 928 | |||
XM_011538803.1 | XP_011537105.1 | 2 | 933 | |||
XM_011538804.2 | XP_011537106.1 | 2 | 1141 | |||
XM_011538805.1 | XP_011537107.1 | 2 | 1782 | |||
XM_011538806.2 | XP_011537108.1 | 2 | 1081 | |||
XM_017020009.1 | XP_016875498.1 | 2 | 1076 | |||
XM_017020010.1 | XP_016875499.1 | 2 | 928 | |||
XM_017020011.1 | XP_016875500.1 | 3 | 954 | |||
XM_017020012.1 | XP_016875501.1 | 2 | 1081 | |||
GenBank mRNA | AK315697.1 | - | 2 | 1090 | ||
AL136825.1 | - | 2 | 1048 | |||
BC030704.1 | - | 2 | 923 | 4825887 |