Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032214.3 | NP_115590.1 | 8 | 1668 | ||
NM_175077.2 | NP_778252.1 | 8 | 1618 | |||
GenBank mRNA | AK025645.1 | - | 8 | 1305 | ||
AK291513.1 | - | 8 | 1648 | |||
BC042041.1 | - | 8 | 1607 | 4429896 |