Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYND12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146192.1 | NP_001139664.1 | 7 | 1381 | ||
NM_032257.4 | NP_115633.3 | 8 | 1553 | |||
GenBank mRNA | AK057384.1 | - | 8 | 1553 | ||
AK301839.1 | - | 7 | 1367 | |||
AL136858.1 | - | 8 | 1360 | |||
BC024186.2 | - | 8 | 1542 | 4826255 |