Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317997.1 | NP_001304926.1 | 5 | 840 | ||
NM_032307.4 | NP_115683.3 | 4 | 1238 | |||
GenBank mRNA | AK090882.1 | - | 5 | 840 | ||
BC004407.1 | - | 4 | 1213 | |||
BC137461.1 | - | 4 | 1115 | 9021461 | ||
BC137462.1 | - | 4 | 1115 | 9021462 |