Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NICN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032316.3 | NP_115692.1 | 6 | 2495 | ||
GenBank mRNA | AF538150.1 | - | 6 | 2244 | ||
AK094248.1 | - | 7 | 2200 | |||
BC005050.1 | - | 7 | 1428 | 2820022 | ||
BC050653.1 | - | 7 | 1803 | 6015080 |