Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ING5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032329.4 | NP_115705.2 | 8 | 4801 | ||
XM_017005097.1 | XP_016860586.1 | 8 | 5869 | |||
XM_017005098.1 | XP_016860587.1 | 9 | 5918 | |||
GenBank mRNA | AK128322.1 | - | 1 | 2742 |