Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRMS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032352.3 | NP_115728.2 | 4 | 509 | ||
XM_005268128.1 | XP_005268185.1 | 4 | 582 | |||
XM_006720274.2 | XP_006720337.1 | 5 | 572 | |||
XM_006720275.2 | XP_006720338.1 | 5 | 572 | |||
XM_017021703.1 | XP_016877192.1 | 4 | 329 | |||
XM_017021704.1 | XP_016877193.1 | 5 | 483 | |||
XM_017021705.1 | XP_016877194.1 | 4 | 406 | |||
GenBank mRNA | AK096496.1 | - | 3 | 232 | ||
AK297106.1 | - | 5 | 572 | |||
AK309431.1 | - | 2 | 127 | |||
AK310359.1 | - | 4 | 443 | |||
AK315424.1 | - | 4 | 440 | |||
AY827074.1 | - | 4 | 495 | |||
BC006250.2 | - | 4 | 489 | 3947694 | ||
BU507838.1 | - | 4 | 553 |