Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF43 SIRNA Assays ›
Gene Symbol:
ZNF43
Gene Name:
zinc finger protein 43
Gene Aliases:
HTF6, KOX27, ZNF39L1
Chromosome Location:
Chr.19: 21804946 - 21852095 on Build GRCh38
UniGene ID:
Hs.534365
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
13132
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF43
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256648.1
NP_001243577.1
7
1076
NM_001256649.1
NP_001243578.1
7
941
NM_001256650.1
NP_001243579.1
6
813
NM_001256651.1
NP_001243580.1
3
427
NM_001256653.1
NP_001243582.1
4
554
NM_001256654.1
NP_001243583.1
3
436
NM_003423.3
NP_003414.2
4
563
XM_011528257.2
XP_011526559.1
4
485
XM_011528259.2
XP_011526561.1
4
1042
XM_017027207.1
XP_016882696.1
7
6554
XM_017027208.1
XP_016882697.1
7
1234
XM_017027209.1
XP_016882698.1
7
6287
XM_017027210.1
XP_016882699.1
6
6078
XM_017027211.1
XP_016882700.1
7
6221
XM_017027212.1
XP_016882701.1
6
1081
XM_017027213.1
XP_016882702.1
7
6329
XM_017027214.1
XP_016882703.1
5
1004
XM_017027215.1
XP_016882704.1
6
843
XM_017027216.1
XP_016882705.1
5
629
GenBank mRNA
AK291353.1
-
7
891
BC006528.2
-
4
530
3687640
BT009756.1
-
3
375
BX471947.1
-
4
564
CR933640.1
-
7
1036
DA784080.1
-
4
559
DB158705.1
-
3
415
DR002502.1
-
4
485
M61869.1
-
3
390
X59244.1
-
6
712
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0